Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DACT3
Entrez Gene ID
612178
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
-
Chromosome Location
Chr.1: 109247743 - 109264655 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116573.2 XP_013972048.2 2 - 3 1325 122

View Full Product Details