Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WBP1
Entrez Gene ID
475787
Gene Name
WW domain binding protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 48671039 - 48673817 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847547.5 XP_852640.1 2 - 3 670 71

View Full Product Details