Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CINP
Entrez Gene ID
612609
Gene Name
cyclin dependent kinase 2 interacting protein
Gene Aliases
-
Chromosome Location
Chr.8: 70307464 - 70332838 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850344.5 XP_855437.2 2 - 3 195 150

View Full Product Details