Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BOLA3
Entrez Gene ID
612287
Gene Name
bolA family member 3
Gene Aliases
-
Chromosome Location
Chr.17: 48941730 - 48952327 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850028.5 XP_855121.1 3 - 4 325 65

View Full Product Details