Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHE
Entrez Gene ID
612268
Gene Name
Src homology 2 domain containing E
Gene Aliases
-
Chromosome Location
Chr.7: 42853116 - 42869293 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850008.5 XP_855101.1 4 - 5 1288 102

View Full Product Details