Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCFC1R1
Entrez Gene ID
610727
Gene Name
host cell factor C1 regulator 1
Gene Aliases
-
Chromosome Location
Chr.6: 38164215 - 38165921 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014114672.2 XP_013970147.1 2 - 3 246 127

View Full Product Details