Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPLX4
Entrez Gene ID
612922
Gene Name
complexin 4
Gene Aliases
-
Chromosome Location
Chr.1: 16900078 - 16922914 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847542.5 XP_852635.1 - 1033 105

View Full Product Details