Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELENOM
Entrez Gene ID
611981
Gene Name
selenoprotein M
Gene Aliases
SELM
Chromosome Location
Chr.26: 24108342 - 24111005 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115014.2 NP_001108486.1 3 - 4 252 71
GenBank mRNA DN878810.1 - 3 - 4 131 71

View Full Product Details