Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GXYLT2
Entrez Gene ID
610815
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.20: 19184892 - 19253144 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022407337.1 XP_022263045.1 - 333 80

View Full Product Details