Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEFL
Entrez Gene ID
100856220
Gene Name
neurofilament light
Gene Aliases
-
Chromosome Location
Chr.25: 32463361 - 32468965 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639880.4 XP_003639928.1 1 - 2 1178 84

View Full Product Details