Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPHS1
Entrez Gene ID
477999
Gene Name
selenophosphate synthetase 1
Gene Aliases
-
Chromosome Location
Chr.2: 23147303 - 23178808 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190410.1 NP_001177339.1 - 273 64
XM_014122200.2 XP_013977675.1 - 408 64

View Full Product Details