Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRIP1
Entrez Gene ID
612713
Gene Name
cysteine rich protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 72784726 - 72786108 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850438.5 XP_855531.1 3 - 4 304 78

View Full Product Details