Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF5A2
Entrez Gene ID
488164
Gene Name
eukaryotic translation initiation factor 5A2
Gene Aliases
-
Chromosome Location
Chr.34: 35309812 - 35322382 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022414133.1 XP_022269841.1 3 - 4 447 114

View Full Product Details