Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORF4L2
Entrez Gene ID
481001
Gene Name
mortality factor 4 like 2
Gene Aliases
-
Chromosome Location
Chr.X: 77084940 - 77097900 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003435631.4 XP_003435679.1 1 - 2 179 104

View Full Product Details