Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX3X
Entrez Gene ID
480886
Gene Name
DEAD-box helicase 3 X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 35759367 - 35789759 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641254.3 XP_005641311.1 5 - 6 570 89
XM_005641255.2 XP_005641312.1 5 - 6 573 89
XM_005641256.3 XP_005641313.1 5 - 6 572 89
XM_856175.5 XP_861268.1 5 - 6 568 89

View Full Product Details