Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH1
Entrez Gene ID
487001
Gene Name
junctophilin 1
Gene Aliases
-
Chromosome Location
Chr.29: 22701531 - 22785712 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005638053.1 XP_005638110.1 4 - 5 2211 75
XM_005638054.1 XP_005638111.1 4 - 5 2211 75
XM_022412295.1 XP_022268003.1 4 - 5 2211 75
XM_544130.4 XP_544130.3 4 - 5 2211 75
XM_844878.3 XP_849971.1 4 - 5 2211 75

View Full Product Details