Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH1
Entrez Gene ID
487001
Gene Name
junctophilin 1
Gene Aliases
-
Chromosome Location
Chr.29: 22701531 - 22785712 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844878.3 XP_849971.1 5 - 6 2305 103

View Full Product Details