Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT5
Entrez Gene ID
100687760
Gene Name
keratin 5
Gene Aliases
-
Chromosome Location
Chr.27: 2567178 - 2573543 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001346035.1 NP_001332964.1 8 - 9 1588 68

View Full Product Details