Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOD
Entrez Gene ID
478604
Gene Name
apolipoprotein D
Gene Aliases
-
Chromosome Location
Chr.33: 30490149 - 30504804 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014110289.1 XP_013965764.1 4 - 5 582 61

View Full Product Details