Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGAM1
Entrez Gene ID
477786
Gene Name
phosphoglycerate mutase 1
Gene Aliases
-
Chromosome Location
Chr.28: 10720235 - 10727709 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_854945.5 XP_860038.2 3 - 4 684 78

View Full Product Details