Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRFAP1
Entrez Gene ID
608548
Gene Name
Morf4 family associated protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 58760074 - 58762225 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005618442.3 XP_005618499.1 2 - 3 561 67
XM_005618443.3 XP_005618500.1 2 - 3 559 67

View Full Product Details