Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDELR2
Entrez Gene ID
612839
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Gene Aliases
-
Chromosome Location
Chr.6: 11796586 - 11815765 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845336.5 XP_850429.1 - 194 76
XM_856309.5 XP_861402.1 - 204 76

View Full Product Details