Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- KDELR2
- Entrez Gene ID
-
612839
- Gene Name
- KDEL endoplasmic reticulum protein retention receptor 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 11796586 - 11815765 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_845336.5 |
XP_850429.1 |
- |
453 |
84 |
View Full Product Details