Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A2
Entrez Gene ID
403448
Gene Name
solute carrier family 10 member 2
Gene Aliases
-
Chromosome Location
Chr.22: 52523679 - 52541976 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002968.1 NP_001002968.1 5 - 6 922 91
GenBank mRNA AJ581082.1 - 5 - 6 922 91

View Full Product Details