Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
NKIRAS2
Entrez Gene ID
480518
Gene Name
NFKB inhibitor interacting Ras like 2
Gene Aliases
-
Chromosome Location
Chr.9: 20851014 - 20854953 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537638.5 XP_537638.1 - 227 129

View Full Product Details