Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMA8
Entrez Gene ID
100856395
Gene Name
proteasome 20S subunit alpha 8
Gene Aliases
-
Chromosome Location
Chr.7: 62432477 - 62472984 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639174.3 XP_003639222.1 4 - 5 578 113
XM_022421515.1 XP_022277223.1 4 - 5 579 113

View Full Product Details