Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKOR1
Entrez Gene ID
610933
Gene Name
SKI family transcriptional corepressor 1
Gene Aliases
LBXCOR1
Chromosome Location
Chr.30: 31935860 - 31948161 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022412722.1 XP_022268430.1 9 - 10 3243 127

View Full Product Details