Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPPED1
Entrez Gene ID
609458
Gene Name
metallophosphoesterase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 22075534 - 22155974 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846716.5 XP_851809.3 2 - 3 434 87

View Full Product Details