Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABPB1
Entrez Gene ID
608465
Gene Name
GA binding protein transcription factor subunit beta 1
Gene Aliases
GABPAP
Chromosome Location
Chr.30: 16239321 - 16303701 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005638401.3 XP_005638458.1 3 - 4 770 138
XM_022412503.1 XP_022268211.1 2 - 3 280 138
XM_022412505.1 XP_022268213.1 3 - 4 305 138
XM_022412506.1 XP_022268214.1 2 - 3 280 138
XM_022412507.1 XP_022268215.1 2 - 3 280 138
XM_022412508.1 XP_022268216.1 2 - 3 280 138
XM_845774.5 XP_850867.1 3 - 4 447 138

View Full Product Details