Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THRA
Entrez Gene ID
403601
Gene Name
thyroid hormone receptor alpha
Gene Aliases
-
Chromosome Location
Chr.9: 22469466 - 22493445 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286862.1 NP_001273791.1 - 1264 75

View Full Product Details