Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPB7
Entrez Gene ID
487423
Gene Name
heat shock protein family B (small) member 7
Gene Aliases
-
Chromosome Location
Chr.2: 81628051 - 81631939 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_859936.4 XP_865029.1 2 - 3 441 76

View Full Product Details