Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
490436
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.7: 42457294 - 42466549 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022421310.1 XP_022277018.1 9 - 9 1580 100
XM_022421311.1 XP_022277019.1 8 - 8 1515 100
XM_022421313.1 XP_022277021.1 10 - 10 1493 100
XM_022421314.1 XP_022277022.1 9 - 9 1580 100
XM_022421315.1 XP_022277023.1 8 - 8 1410 100
XM_022421316.1 XP_022277024.1 8 - 8 1390 100
XM_022421317.1 XP_022277025.1 9 - 9 1580 100

View Full Product Details