Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR14L
Entrez Gene ID
477545
Gene Name
proline rich 14 like
Gene Aliases
-
Chromosome Location
Chr.26: 24570777 - 24624151 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636489.3 XP_005636546.1 5 - 5 5926 123
XM_014107906.2 XP_013963381.1 4 - 4 5399 123
XM_014107908.2 XP_013963383.1 3 - 3 5276 123
XM_022410718.1 XP_022266426.1 5 - 5 5859 123
XM_022410719.1 XP_022266427.1 5 - 5 5841 123
XM_022410720.1 XP_022266428.1 4 - 4 5743 123
XM_022410721.1 XP_022266429.1 4 - 4 5725 123
XM_534740.6 XP_534740.2 4 - 4 5804 123

View Full Product Details