Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADRA1A
Entrez Gene ID
403866
Gene Name
adrenoceptor alpha 1A
Gene Aliases
ADRA1C, ALPHA-1A
Chromosome Location
Chr.25: 30690959 - 30801484 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195843.1 NP_001182772.1 1 - 1 800 126
XM_005635607.3 XP_005635664.1 3 - 3 1857 126
XM_022409542.1 XP_022265250.1 2 - 2 2107 126
XM_022409544.1 XP_022265252.1 2 - 2 2111 126
XM_022409545.1 XP_022265253.1 2 - 2 2109 126
XM_022409546.1 XP_022265254.1 2 - 2 2118 126
GenBank mRNA AF068283.1 - 1 - 1 801 126

View Full Product Details