Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR7
Entrez Gene ID
611253
Gene Name
proline rich 7, synaptic
Gene Aliases
-
Chromosome Location
Chr.4: 35913300 - 35923823 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014112979.2 XP_013968454.1 2 - 2 2616 87
XM_848895.5 XP_853988.1 3 - 3 875 87

View Full Product Details