Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGLN1
Entrez Gene ID
488971
Gene Name
egl-9 family hypoxia inducible factor 1
Gene Aliases
-
Chromosome Location
Chr.4: 8053486 - 8110791 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546089.5 XP_546089.3 - 890 135

View Full Product Details