Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJA3
Entrez Gene ID
479865
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
-
Chromosome Location
Chr.6: 36921941 - 36967718 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014114657.2 XP_013970132.1 7 - 8 1041 146
XM_536990.6 XP_536990.2 7 - 8 1041 146
XM_846658.5 XP_851751.1 7 - 8 1042 146

View Full Product Details