Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBF4B
Entrez Gene ID
480497
Gene Name
DBF4 zinc finger B
Gene Aliases
-
Chromosome Location
Chr.9: 18734678 - 18768128 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022422894.1 XP_022278602.1 - 484 109

View Full Product Details