Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLF4
Entrez Gene ID
481657
Gene Name
Kruppel like factor 4
Gene Aliases
-
Chromosome Location
Chr.11: 62881840 - 62886676 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005626996.2 XP_005627053.1 3 - 3 1124 66

View Full Product Details