Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MORF4L2
Entrez Gene ID
481001
Gene Name
mortality factor 4 like 2
Gene Aliases
-
Chromosome Location
Chr.X: 77084940 - 77097900 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003435631.4 XP_003435679.1 4 - 4 912 164
XM_003435632.4 XP_003435680.1 4 - 4 861 164

View Full Product Details