Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYLS1
Entrez Gene ID
609650
Gene Name
HYLS1 centriolar and ciliogenesis associated
Gene Aliases
-
Chromosome Location
Chr.5: 8589606 - 8605196 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619583.2 XP_005619640.1 1 - 1 1017 136
XM_005619584.2 XP_005619641.1 1 - 1 790 136
XM_005619585.3 XP_005619642.1 1 - 1 875 136
XM_005619586.2 XP_005619643.1 1 - 1 732 136
XM_014113310.2 XP_013968785.1 1 - 1 1010 136
XM_014113311.2 XP_013968786.1 1 - 1 878 136
XM_022418111.1 XP_022273819.1 1 - 1 997 136
XM_022418112.1 XP_022273820.1 1 - 1 879 136
XM_846949.5 XP_852042.1 1 - 1 757 136

View Full Product Details