Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCH1
Entrez Gene ID
609393
Gene Name
GTP cyclohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.8: 30733422 - 30784067 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005623417.3 XP_005623474.1 6 - 6 847 66

View Full Product Details