Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD11B2
Entrez Gene ID
489758
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.5: 81966397 - 81970301 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620822.3 XP_005620879.1 4 - 4 2745 82
GenBank mRNA DQ195098.1 - 4 - 4 674 82

View Full Product Details