Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNJ11
Entrez Gene ID
485401
Gene Name
potassium inwardly rectifying channel subfamily J member 11
Gene Aliases
Kir6.2
Chromosome Location
Chr.21: 39953461 - 39957615 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005633719.3 XP_005633776.1 2 - 2 681 145

View Full Product Details