Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COR5M13P
Entrez Gene ID
483519
Gene Name
cOR5M13P olfactory receptor family 5 subfamily M-like
Gene Aliases
OR5M11, cOR5M16
Chromosome Location
Chr.18: 39412932 - 39425375 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_540638.4 XP_540638.4 2 - 2 1043 189

View Full Product Details