Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
KLF6
Entrez Gene ID
487151
Gene Name
Kruppel like factor 6
Gene Aliases
-
Chromosome Location
Chr.2: 31654142 - 31664009 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544279.6 XP_544279.3 - 456 177

View Full Product Details