Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP13
Entrez Gene ID
489054
Gene Name
dual specificity phosphatase 13
Gene Aliases
-
Chromosome Location
Chr.4: 25360291 - 25373299 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_859212.4 XP_864305.1 1 - 2 215 125

View Full Product Details