Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
RTN2
Entrez Gene ID
484444
Gene Name
reticulon 2
Gene Aliases
OPA3
Chromosome Location
Chr.1: 109976820 - 110080252 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_850003.5 XP_855096.1 2 - 2 275 133

View Full Product Details