Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
BMP4
Entrez Gene ID
490695
Gene Name
bone morphogenetic protein 4
Gene Aliases
-
Chromosome Location
Chr.8: 29997563 - 30254307 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287170.1 NP_001274099.1 3 - 3 685 151
XM_022421759.1 XP_022277467.1 3 - 3 529 151
XM_022421760.1 XP_022277468.1 5 - 5 850 151
XM_022421761.1 XP_022277469.1 5 - 5 786 151
XM_022421762.1 XP_022277470.1 6 - 6 984 151
XM_022421763.1 XP_022277471.1 5 - 5 773 151
XM_022421764.1 XP_022277472.1 7 - 7 1023 151

View Full Product Details