Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXD10
Entrez Gene ID
609973
Gene Name
homeobox D10
Gene Aliases
-
Chromosome Location
Chr.36: 19924352 - 19927706 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005640343.2 XP_005640400.1 1 - 1 804 132

View Full Product Details